siRNA Details
| Gene Symbol: Chek2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016681.3 | NP_057890.1 | 12, 13 | 1432 | ||
| GenBank mRNA | AF086905.1 | - | 11, 12 | 1393 | ||
| AK044913.1 | - | 12, 13 | 1433 | |||
| AK170235.1 | - | 11, 12 | 1376 | |||
| BC056617.1 | - | 12, 13 | 1456 | 4977960 | ||

Chek2
Genomic Map