siRNA Details
| Gene Symbol: Grb14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016719.1 | NP_057928.1 | 5 | 760 | ||
| GenBank mRNA | AF155647.1 | - | 5 | 760 | ||
| AK010849.1 | - | 5 | 777 | |||
| AK010903.1 | - | 5 | 777 | |||
| AK087758.1 | - | 3 | 2390 | |||
| AK147041.1 | - | 5 | 769 | |||
| BC138581.1 | - | 5 | 666 | 8861601 | ||
| BC145913.1 | - | 5 | 666 | 40131216 | ||

Grb14
Genomic Map