siRNA Details
| Gene Symbol: Tnfsf14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019418.3 | NP_062291.1 | 4 | 637 | ||
| GenBank mRNA | AB029155.1 | - | 4 | 648 | ||
| AF123385.1 | - | 4 | 525 | |||
| AF227533.1 | - | 4 | 535 | |||
| BC125477.1 | - | 4 | 614 | 40129992 | ||
| BC125479.1 | - | 4 | 614 | 40129994 | ||

Tnfsf14
Genomic Map