siRNA Details
Gene Symbol: Slc7a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016972.2 | NP_058668.1 | 7 | 1508 | ||
GenBank mRNA | AF171668.1 | - | 7 | 1163 | ||
BC059004.1 | - | 7 | 1508 | 6849059 | ||
Y19022.1 | - | 7 | 1175 |
