siRNA Details
| Gene Symbol: Srpx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016911.4 | NP_058607.1 | 8 | 1111 | ||
| GenBank mRNA | AB028049.1 | - | 8 | 1038 | ||
| AB028050.1 | - | 6 | 786 | |||
| AK034696.1 | - | 8 | 1113 | |||
| AK159662.1 | - | 7 | 1099 | |||
| BC022572.1 | - | 8 | 1067 | 3993025 | ||

Srpx
Genomic Map