siRNA Details
| Gene Symbol: Hnrnpu | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016805.2 | NP_058085.2 | 7 | 1424 | ||
| GenBank mRNA | AF073992.1 | - | 7 | 1361 | ||
| AK029944.1 | - | 7 | 1370 | |||
| AK037517.1 | - | 7 | 1366 | |||
| AK089058.1 | - | 7 | 1425 | |||
| AK145499.1 | - | 7 | 1427 | |||
| AK145682.1 | - | 7 | 1427 | |||
| AK159084.1 | - | 7 | 1393 | |||
| AK159614.1 | - | 7 | 1429 | |||
| AK159956.1 | - | 7 | 1366 | |||
| AK162264.1 | - | 7 | 1429 | |||
| AK165844.1 | - | 7 | 1424 | |||
| AK167200.1 | - | 7 | 1427 | |||
| AK168657.1 | - | 7 | 1366 | |||
| AK190289.1 | - | 4 | 342 | |||
| BC018353.1 | - | 7 | 1269 | 3584692 | ||

Hnrnpu
Genomic Map