siRNA Details
Gene Symbol: Hnrnpu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016805.2 | NP_058085.2 | 5 | 1219 | ||
GenBank mRNA | AF073992.1 | - | 5 | 1156 | ||
AK029944.1 | - | 5 | 1165 | |||
AK037517.1 | - | 5 | 1161 | |||
AK089058.1 | - | 5 | 1220 | |||
AK145499.1 | - | 5 | 1222 | |||
AK145682.1 | - | 5 | 1221 | |||
AK159084.1 | - | 5 | 1188 | |||
AK159614.1 | - | 5 | 1224 | |||
AK159956.1 | - | 5 | 1161 | |||
AK162264.1 | - | 5 | 1224 | |||
AK165844.1 | - | 5 | 1219 | |||
AK167200.1 | - | 5 | 1221 | |||
AK168657.1 | - | 5 | 1161 | |||
AK190289.1 | - | 2 | 137 | |||
BC018353.1 | - | 5 | 1064 | 3584692 |
