siRNA Details
| Gene Symbol: Rif1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175238.5 | NP_780447.4 | 19, 20 | 2159 | ||
| GenBank mRNA | AK088688.1 | - | 7, 8 | 1851 | ||
| AY428571.2 | - | 19, 20 | 2143 | |||
| AY585206.1 | - | 18, 19 | 2152 | |||
| JX255737.1 | - | 18, 19 | 2095 | |||

Rif1
Genomic Map