siRNA Details
| Gene Symbol: Fubp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057172.3 | NP_476513.2 | 11, 12 | 1066 | ||
| GenBank mRNA | AK028418.1 | - | 11, 12 | 1067 | ||
| AK032517.1 | - | 11, 12 | 1033 | |||
| AK138011.1 | - | 11, 12 | 1011 | |||
| AK160815.1 | - | 11, 12 | 1004 | |||
| AK166066.1 | - | 11, 12 | 1011 | |||
| AK167730.1 | - | 11, 12 | 1046 | |||
| BC014763.1 | - | 11, 12 | 1029 | 4511225 | ||
| BC023780.1 | - | 11, 12 | 1047 | 5347067 | ||

Fubp1
Genomic Map