siRNA Details
| Gene Symbol: Rnf24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178607.4 | NP_848722.1 | 5, 6 | 469 | ||
| GenBank mRNA | AK015699.1 | - | 6, 7 | 529 | ||
| AK048813.1 | - | 5, 6 | 427 | |||
| AK083665.1 | - | 5, 6 | 459 | |||
| AK089207.1 | - | 5, 6 | 456 | |||
| AK154093.1 | - | 5, 6 | 406 | |||
| AK165427.1 | - | 5, 6 | 427 | |||
| BC096529.1 | - | 5, 6 | 402 | 6406985 | ||

Rnf24
Genomic Map