siRNA Details
| Gene Symbol: Ccdc39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026222.2 | NP_080498.1 | 7 | 901 | ||
| GenBank mRNA | AK014839.1 | - | 7 | 893 | ||
| AK029948.1 | - | 7 | 857 | |||
| AK030087.1 | - | 7 | 868 | |||
| AK083226.1 | - | 7 | 773 | |||
| BC138917.1 | - | 7 | 863 | 8861939 | ||
| BC138920.1 | - | 7 | 863 | 8861942 | ||

Ccdc39
Genomic Map