siRNA Details
| Gene Symbol: Umodl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177465.4 | NP_803416.2 | 21 | 4024 | ||
| GenBank mRNA | AK040446.1 | - | 13 | 2525 | ||
| AY771616.1 | - | 22 | 3941 | |||
| AY771617.1 | - | 21 | 4028 | |||
| AY771618.1 | - | 21 | 3683 | |||
| BC051427.1 | - | 21 | 3679 | |||
| BC132381.1 | - | 8 | 1213 | |||

Umodl1
Genomic Map