siRNA Details
| Gene Symbol: Pibf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029320.3 | NP_083596.2 | 2 | 791 | ||
| GenBank mRNA | AK015781.1 | - | 2 | 258 | ||
| AK017107.1 | - | 2 | 792 | |||
| AK030385.1 | - | 2 | 272 | |||
| AK045133.1 | - | 2 | 286 | |||
| AK050974.1 | - | 1 | 1158 | |||
| BC113804.1 | - | 2 | 268 | |||
| BC129811.1 | - | 2 | 268 | 40062382 | ||
| BC129812.1 | - | 2 | 268 | 40062383 | ||

Pibf1
Genomic Map