siRNA Details
| Gene Symbol: Ppp6r3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164159.1 | NP_001157631.1 | 10 | 1371 | ||
| NM_028999.1 | NP_083275.1 | 11 | 1519 | |||
| NM_029456.2 | NP_083732.1 | 11 | 1519 | |||
| GenBank mRNA | AK018652.1 | - | 10 | 1360 | ||
| AK032179.1 | - | 2 | 290 | |||
| AK035236.1 | - | 11 | 1499 | |||
| AK088526.1 | - | 10 | 1339 | |||
| AK129392.1 | - | 11 | 1461 | |||
| BC008529.1 | - | 11 | 1502 | 3601021 | ||
| CF727136.1 | - | 1 | 36 | |||

Ppp6r3
Genomic Map