siRNA Details
| Gene Symbol: Ppp1r10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163818.1 | NP_001157290.1 | 3, 4 | 717 | ||
| GenBank mRNA | AJ504718.1 | - | 3, 4 | 723 | ||
| AK053183.1 | - | 4, 5 | 736 | |||
| AK145485.1 | - | 2, 3 | 161 | |||
| BC030903.1 | - | 3, 4 | 724 | |||
| BC052059.1 | - | 3, 4 | 720 | 5702171 | ||

Ppp1r10
Genomic Map