siRNA Details
| Gene Symbol: Ppp1r10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163818.1 | NP_001157290.1 | 13 | 1844 | ||
| GenBank mRNA | AJ504718.1 | - | 13 | 1850 | ||
| AK145485.1 | - | 12 | 1288 | |||
| BC029765.1 | - | 5 | 518 | |||
| BC052059.1 | - | 13 | 1847 | 5702171 | ||

Ppp1r10
Genomic Map