siRNA Details
| Gene Symbol: Ppp1r10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163818.1 | NP_001157290.1 | 6, 7 | 988 | ||
| GenBank mRNA | AJ504718.1 | - | 6, 7 | 994 | ||
| AK053183.1 | - | 7, 8 | 1007 | |||
| AK145485.1 | - | 5, 6 | 432 | |||
| BC030903.1 | - | 6, 7 | 995 | |||
| BC052059.1 | - | 6, 7 | 991 | 5702171 | ||

Ppp1r10
Genomic Map