siRNA Details
| Gene Symbol: Tmem38b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028053.2 | NP_082329.1 | 5 | 709 | ||
| GenBank mRNA | AB261159.1 | - | 5 | 645 | ||
| AK005479.1 | - | 5 | 692 | |||
| AK136131.1 | - | 4 | 459 | |||
| AK150377.1 | - | 5 | 687 | |||
| AK150892.1 | - | 5 | 687 | |||
| AK153075.1 | - | 5 | 674 | |||
| BC011072.1 | - | 5 | 687 | 3978742 | ||

Tmem38b
Genomic Map