siRNA Details
| Gene Symbol: Rragd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027491.2 | NP_081767.2 | 4 | 1201 | ||
| GenBank mRNA | AK017818.1 | - | 3 | 561 | ||
| BC038137.1 | - | 4 | 690 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rragd| Gene Symbol: Rragd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027491.2 | NP_081767.2 | 4 | 1201 | ||
| GenBank mRNA | AK017818.1 | - | 3 | 561 | ||
| BC038137.1 | - | 4 | 690 | |||