siRNA Details
| Gene Symbol: Stbd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175096.3 | NP_780305.1 | 2 | 970 | ||
| GenBank mRNA | AK050416.1 | - | 2 | 971 | ||
| BC052666.1 | - | 2 | 931 | |||
| BC118615.1 | - | 3 | 1039 | 40107322 | ||
| BC118661.1 | - | 2 | 953 | 40107321 | ||

Stbd1
Genomic Map