siRNA Details
| Gene Symbol: Zfp644 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026856.2 | NP_081132.2 | 3 | 1975 | ||
| GenBank mRNA | AK122471.1 | - | 1 | 1455 | ||
| AK144865.1 | - | 6 | 2151 | |||
| BC057405.1 | - | 3 | 1854 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp644| Gene Symbol: Zfp644 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026856.2 | NP_081132.2 | 3 | 1975 | ||
| GenBank mRNA | AK122471.1 | - | 1 | 1455 | ||
| AK144865.1 | - | 6 | 2151 | |||
| BC057405.1 | - | 3 | 1854 | |||