siRNA Details
| Gene Symbol: Zfp644 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026856.2 | NP_081132.2 | 3 | 620 | ||
| GenBank mRNA | AK053146.1 | - | 3 | 621 | ||
| AK122471.1 | - | 1 | 100 | |||
| AK144865.1 | - | 6 | 796 | |||
| AK162026.1 | - | 3 | 654 | |||
| BB663850.1 | - | 3 | 621 | |||
| BC057405.1 | - | 3 | 499 | |||

Zfp644
Genomic Map