siRNA Details
| Gene Symbol: Slc46a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026740.2 | NP_081016.2 | 1, 2 | 312 | ||
| GenBank mRNA | AK003278.1 | - | 1, 2 | 295 | ||
| AK170505.1 | - | 1, 2 | 305 | |||
| AK220201.1 | - | 1, 2 | 272 | |||
| BC057976.1 | - | 1, 2 | 297 | 5052851 | ||
| CX219335.1 | - | 1, 2 | 312 | |||

Slc46a1
Genomic Map