siRNA Details
Gene Symbol: Zfp622 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144523.2 | NP_653106.1 | 5 | 1371 | ||
GenBank mRNA | AK004048.1 | - | 1 | 1100 | ||
AK031232.1 | - | 5 | 1371 | |||
AK149106.1 | - | 5 | 1302 | |||
AK154951.1 | - | 5 | 1297 | |||
BC006964.1 | - | 5 | 1276 | 3153893 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001009652, XM_008760822, BC088214.
Panther Classification:
Gene Ontology Categories:
