siRNA Details
| Gene Symbol: Ldlrad4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172631.3 | NP_766219.2 | 6 | 1215 | ||
| GenBank mRNA | AK040289.1 | - | 4 | 624 | ||
| AK052338.1 | - | 6 | 1216 | |||
| BC022716.1 | - | 3 | 336 | |||
| BC096371.1 | - | 5 | 976 | 3583946 | ||
| BC131933.1 | - | 5 | 823 | 40130210 | ||
| BC131935.1 | - | 5 | 823 | 40130212 | ||

Ldlrad4
Genomic Map