siRNA Details
Gene Symbol: Setd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028262.3 | NP_082538.2 | 11 | 1276 | ||
GenBank mRNA | AK011993.1 | - | 11 | 1256 | ||
AK031017.1 | - | 3 | 1839 | |||
AK031371.1 | - | 11 | 1239 | |||
AK146777.1 | - | 11 | 1270 | |||
AK166570.1 | - | 11 | 1208 | |||
AY513271.1 | - | 11 | 1209 | |||
BC016123.1 | - | 11 | 1206 | 4503842 | ||
BC019973.1 | - | 11 | 1206 | 4195130 | ||
BC057968.1 | - | 8 | 896 | 4979330 | ||
BC108358.1 | - | 11 | 1247 |
