siRNA Details
| Gene Symbol: Setd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028262.3 | NP_082538.2 | 8, 9 | 988 | ||
| GenBank mRNA | AK011993.1 | - | 8, 9 | 968 | ||
| AK031371.1 | - | 8, 9 | 951 | |||
| AK146777.1 | - | 8, 9 | 982 | |||
| AK166570.1 | - | 8, 9 | 920 | |||
| AY513271.1 | - | 8, 9 | 921 | |||
| BC016123.1 | - | 8, 9 | 918 | 4503842 | ||
| BC019973.1 | - | 8, 9 | 918 | 4195130 | ||
| BC108358.1 | - | 8, 9 | 959 | |||

Setd3
Genomic Map