siRNA Details
| Gene Symbol: Krr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178610.4 | NP_848725.2 | 5, 6 | 692 | ||
| GenBank mRNA | AK011972.1 | - | 5, 6 | 602 | ||
| AK012108.1 | - | 5, 6 | 630 | |||
| AK030560.1 | - | 5, 6 | 604 | |||
| AK032472.1 | - | 5, 6 | 604 | |||
| AK036447.1 | - | 4, 5 | 690 | |||
| AK041854.1 | - | 5, 6 | 604 | |||
| AK146926.1 | - | 5, 6 | 610 | |||
| AK168675.1 | - | 5, 6 | 604 | |||
| BC089510.1 | - | 5, 6 | 609 | 30246541 | ||
| BC094236.1 | - | 5, 6 | 593 | |||

Krr1
Genomic Map