siRNA Details
| Gene Symbol: Zfp410 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144833.3 | NP_659082.1 | 11 | 1785 | ||
| GenBank mRNA | AF295806.1 | - | 11 | 1547 | ||
| AK051939.1 | - | 9 | 1185 | |||
| AK083003.1 | - | 11 | 1528 | |||
| AK132345.1 | - | 11 | 1524 | |||
| AK181763.1 | - | 2 | 195 | |||
| BC021528.1 | - | 11 | 1540 | 5353673 | ||
| BC056957.1 | - | 11 | 1482 | |||

Zfp410
Genomic Map