siRNA Details
| Gene Symbol: Zkscan6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK007308.1 | - | 7 | 1363 | ||
| AK015125.1 | - | 7 | 1336 | |||
| BC043075.1 | - | 8 | 1394 | 6404784 | ||
| BC096688.1 | - | 3 | 449 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zkscan6| Gene Symbol: Zkscan6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK007308.1 | - | 7 | 1363 | ||
| AK015125.1 | - | 7 | 1336 | |||
| BC043075.1 | - | 8 | 1394 | 6404784 | ||
| BC096688.1 | - | 3 | 449 | |||