siRNA Details
| Gene Symbol: Tspyl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029836.3 | NP_084112.1 | 7 | 2104 | ||
| GenBank mRNA | AY273809.1 | - | 7 | 1945 | ||
| BC004006.1 | - | 7 | 1751 | |||
| BC030334.1 | - | 5 | 1251 | |||
| BC043694.1 | - | 7 | 1818 | |||
| BC054393.1 | - | 7 | 2024 | 6491672 | ||

Tspyl2
Genomic Map