siRNA Details
| Gene Symbol: Tspyl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029836.3 | NP_084112.1 | 1 | 494 | ||
| GenBank mRNA | AK018700.1 | - | 1 | 436 | ||
| AY273809.1 | - | 1 | 335 | |||
| BC004006.1 | - | 1 | 142 | |||
| BC043694.1 | - | 1 | 250 | |||
| BC054393.1 | - | 1 | 414 | 6491672 | ||

Tspyl2
Genomic Map