siRNA Details
| Gene Symbol: Ldhd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027570.3 | NP_081846.3 | 5 | 563 | ||
| GenBank mRNA | AK029820.1 | - | 712 | |||
| AY092768.1 | - | 5 | 568 | |||
| BC039155.1 | - | 5 | 559 | 4242682 | ||
| BC055443.1 | - | 5 | 563 | 4235345 | ||

Ldhd
Genomic Map