siRNA Details
| Gene Symbol: Ldhd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027570.3 | NP_081846.3 | 11 | 1332 | ||
| GenBank mRNA | AK014649.1 | - | 11 | 1338 | ||
| AK029820.1 | - | 1746 | ||||
| AK037996.1 | - | 729 | ||||
| BC039155.1 | - | 11 | 1328 | 4242682 | ||
| BC055443.1 | - | 11 | 1332 | 4235345 | ||

Ldhd
Genomic Map