siRNA Details
| Gene Symbol: Rufy3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289776.1 | NP_001276705.1 | 1 | 766 | ||
| NM_001289777.1 | NP_001276706.1 | 2 | 591 | |||
| NM_027530.3 | NP_081806.1 | 1 | 766 | |||
| GenBank mRNA | AK013185.1 | - | 2 | 592 | ||
| AK018184.1 | - | 1 | 744 | |||
| AK045610.1 | - | 1 | 700 | |||
| AK083242.1 | - | 1 | 701 | |||
| BC017648.1 | - | 1 | 155 | 4527858 | ||
| BC049127.1 | - | 1 | 705 | 6821567 | ||
| BC058259.1 | - | 1 | 485 | 6469137 | ||
| CT010404.1 | - | 1 | 139 | |||
| DV068924.1 | - | 2 | 522 | |||

Rufy3
Genomic Map