siRNA Details
| Gene Symbol: Ahi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026203.3 | NP_080479.2 | 15 | 2615 | ||
| GenBank mRNA | AK036336.1 | - | 15 | 2505 | ||
| AY133241.1 | - | 15 | 2508 | |||
| BC038502.1 | - | 8 | 966 | |||
| BC055400.1 | - | 14 | 2337 | 6467369 | ||
| BC065146.1 | - | 15 | 2510 | |||

Ahi1
Genomic Map