siRNA Details
| Gene Symbol: Zmiz2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005867.1 | NP_001005867.1 | 12, 13 | 1820 | ||
| NM_028601.3 | NP_082877.2 | 12, 13 | 1916 | |||
| GenBank mRNA | BC024081.1 | - | 11, 12 | 1666 | ||
| BC034541.1 | - | 2, 3 | 262 | |||
| BC040745.1 | - | 3, 4 | 375 | |||
| BC060652.1 | - | 12, 13 | 1758 | 30354393 | ||
| BC063069.1 | - | 4, 5 | 521 | |||

Zmiz2
Genomic Map