siRNA Details
| Gene Symbol: Zmiz2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005867.1 | NP_001005867.1 | 4, 5 | 487 | ||
| NM_028601.3 | NP_082877.2 | 4, 5 | 583 | |||
| GenBank mRNA | BC024081.1 | - | 3, 4 | 333 | ||
| BC060652.1 | - | 4, 5 | 425 | 30354393 | ||
| CB524312.1 | - | 2, 3 | 146 | |||
| CB527314.1 | - | 3, 4 | 335 | |||

Zmiz2
Genomic Map