siRNA Details
| Gene Symbol: Nub1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF534114.1 | - | 1 | 57 | ||
| AK086301.1 | - | 2 | 267 | |||
| AK134581.1 | - | 2 | 111 | |||
| AK153400.1 | - | 2 | 95 | |||
| BC005640.1 | - | 2 | 96 | |||
| BC018178.1 | - | 2 | 82 | |||
| BC037451.1 | - | 1 | 86 | 4018648 | ||
| U27462.1 | - | 2 | 304 | |||

Nub1
Genomic Map