siRNA Details
| Gene Symbol: Nub1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF534114.1 | - | 9, 10 | 1095 | ||
| AK018166.1 | - | 9, 10 | 1904 | |||
| AK086301.1 | - | 10, 11 | 1305 | |||
| AK134581.1 | - | 10, 11 | 1149 | |||
| AK153400.1 | - | 10, 11 | 1133 | |||
| BC005640.1 | - | 10, 11 | 1134 | |||
| BC018178.1 | - | 10, 11 | 1120 | |||
| BC037451.1 | - | 9, 10 | 1124 | 4018648 | ||
| U27462.1 | - | 10, 11 | 1342 | |||

Nub1
Genomic Map