siRNA Details
| Gene Symbol: Plrg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016784.3 | NP_058064.2 | 10 | 967 | ||
| GenBank mRNA | AF044334.1 | - | 9 | 859 | ||
| AK075993.1 | - | 10 | 955 | |||
| AK076007.1 | - | 10 | 945 | |||
| AK077197.1 | - | 10 | 968 | |||
| AK080068.1 | - | 10 | 974 | |||
| BC006750.1 | - | 10 | 937 | 3709134 | ||
| BC052895.1 | - | 10 | 925 | |||

Plrg1
Genomic Map