siRNA Details
| Gene Symbol: Folh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159706.1 | NP_001153178.1 | 3 | 420 | ||
| NM_016770.3 | NP_058050.3 | 3 | 420 | |||
| GenBank mRNA | AF026380.1 | - | 3 | 402 | ||
| AK002920.1 | - | 3 | 387 | |||
| AK090289.1 | - | 421 | ||||
| BC119604.1 | - | 3 | 364 | 40060385 | ||
| BC119605.1 | - | 3 | 364 | 40060386 | ||

Folh1
Genomic Map