siRNA Details
| Gene Symbol: Chst3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016803.3 | NP_058083.2 | 3 | 691 | ||
| GenBank mRNA | AB008937.1 | - | 2 | 310 | ||
| BC055055.1 | - | 1 | 570 | 6493234 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Chst3| Gene Symbol: Chst3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016803.3 | NP_058083.2 | 3 | 691 | ||
| GenBank mRNA | AB008937.1 | - | 2 | 310 | ||
| BC055055.1 | - | 1 | 570 | 6493234 | ||