siRNA Details
Gene Symbol: Exoc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016857.2 | NP_058553.2 | 13 | 1511 | ||
GenBank mRNA | AF014461.1 | - | 13 | 1446 | ||
AK140450.1 | - | 13 | 1485 | |||
AK162868.1 | - | 11 | 2489 | |||
AK173094.1 | - | 11 | 5998 |
