siRNA Details
Gene Symbol: Bysl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016859.3 | NP_058555.3 | 3, 4 | 885 | ||
GenBank mRNA | AF007802.1 | - | 1, 2 | 114 | ||
AK019239.1 | - | 3, 4 | 620 | |||
AK032993.1 | - | 3, 4 | 617 | |||
AK049195.1 | - | 3, 4 | 878 | |||
AK051704.1 | - | 3, 4 | 875 | |||
AK083033.1 | - | 3, 4 | 872 | |||
AK143027.1 | - | 3, 4 | 886 | |||
AK168301.1 | - | 3, 4 | 623 | |||
BC017530.1 | - | 3, 4 | 612 | 4925307 |
