siRNA Details
| Gene Symbol: Corin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122756.1 | NP_001116228.1 | 3 | 381 | ||
| NM_016869.3 | NP_058565.2 | 3 | 538 | |||
| GenBank mRNA | AB013874.1 | - | 3 | 538 | ||
| BB839745.1 | - | 3 | 381 | |||
| BC093485.1 | - | 3 | 314 | 30648458 | ||
| BC138339.1 | - | 3 | 521 | 8861359 | ||
| BC138340.1 | - | 3 | 521 | 8861360 | ||
| BC145243.1 | - | 3 | 521 | 9053795 | ||

Corin
Genomic Map