siRNA Details
| Gene Symbol: Zpbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001185153.1 | NP_001172082.1 | 4 | 411 | ||
| NM_015785.2 | NP_056600.1 | 4 | 582 | |||
| GenBank mRNA | BC139450.1 | - | 4 | 551 | 8862472 | |
| BC139453.1 | - | 4 | 551 | 8862475 | ||
| BC145628.1 | - | 4 | 380 | 9054196 | ||
| D17569.1 | - | 4 | 571 | |||

Zpbp
Genomic Map