siRNA Details
| Gene Symbol: Map3k6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB021861.4 | - | 10 | 1781 | ||
| BC120565.1 | - | 10 | 1710 | 40129488 | ||
| BC125577.1 | - | 10 | 1710 | 40130092 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Map3k6| Gene Symbol: Map3k6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB021861.4 | - | 10 | 1781 | ||
| BC120565.1 | - | 10 | 1710 | 40129488 | ||
| BC125577.1 | - | 10 | 1710 | 40130092 | ||