siRNA Details
| Gene Symbol: Map3k6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016693.5 | NP_057902.5 | 10, 11 | 1858 | ||
| GenBank mRNA | AB021861.4 | - | 10, 11 | 1856 | ||
| BC120565.1 | - | 10, 11 | 1785 | 40129488 | ||
| BC125577.1 | - | 10, 11 | 1785 | 40130092 | ||

Map3k6
Genomic Map