siRNA Details
| Gene Symbol: Reck | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016678.2 | NP_057887.2 | 9 | 768 | ||
| GenBank mRNA | AB006960.1 | - | 9 | 698 | ||
| AK042266.1 | - | 9 | 769 | |||
| AK143657.1 | - | 9 | 746 | |||
| AK157885.1 | - | 9 | 756 | |||
| BC053025.1 | - | 9 | 707 | |||
| BC132197.1 | - | 9 | 796 | 40130474 | ||
| BC138065.1 | - | 9 | 796 | 8861084 | ||
| BC145133.1 | - | 9 | 797 | |||

Reck
Genomic Map